Anti-FASLG Mouse Monoclonal Antibody (Biotin) [clone: B-R17]

Supplier: Cell Sciences
CDM075
102995-544EA 498.46 USD
102995-544
Anti-FASLG Mouse Monoclonal Antibody (Biotin) [clone: B-R17]
Antibodies
Flow Cytometry: Use 10 ?l to label 10E6 cells or 100 ?l of whole blood. Recognizes the soluble antigen.

Type: Primary
Antigen: CD178
Clonality: Monoclonal
Clone: B-R17
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR