Anti-FAS Mouse Monoclonal Antibody [clone: B-D29]

Supplier: Cell Sciences
CDM070
102995-536EA 553.85 USD
102995-536
Anti-FAS Mouse Monoclonal Antibody [clone: B-D29]
Antibodies
Inhibits FasL induced apoptosis.

Type: Primary
Antigen: CD95
Clonality: Monoclonal
Clone: B-D29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR