Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAL10 Mouse Monoclonal Antibody [clone: B-F42]

Supplier: Cell Sciences
CDM065
102995-526EA 418.46 USD InStock
102995-526
Anti-GAL10 Mouse Monoclonal Antibody [clone: B-F42]
Antibodies
Galectin-10 is an intracellular antigen. Fixation with paraformaldehyde 4% and permeabilization with saponin 0.1% are required before staining. Use 10 µl to label 10E6 cells.

Type: Primary
Antigen: GAL10
Clonality: Monoclonal
Clone: B-F42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty