Would you like to log in again?
POLYTRON® PT 2500 E Homogenizer, Kinematica
Supplier: KINEMATICA INC
11090104
97057-696
POLYTRON® PT 2500 E Homogenizer, Kinematica
The PT2500 E is the ideal solution for cost-conscious laboratories. It is able to perform homogenizations with volumes from 0.05 to 2500 mL.
  • Digital motor speed control from 500 to 30,000 rpm
  • Aggregate tip speeds up to 32 m/s produce the finest particles possible
  • Thermal fuse for overload protection
  • Aggregate replacement using one-handed quick-release coupling
The electronic control system continuously monitors the speed and automatically adjusts it, keeping it constant with changing viscosity
Aggregates with a variety of styles and diameters are available to provide optimum particle size reduction.
The speed is easily seen on the large digital display.
Ordering information: Includes unit cable and stand with vessel holder and safety ring.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty