Would you like to log in again?
Whirl-Pak® Sampling Bags, Standard, Advantec MFS
Supplier: ADVANTEC MFS MS
WP018ZWS
101105-814
Whirl-Pak® Sampling Bags, Standard, Advantec MFS
It is mainly used for sterile sampling of soild and/or liquid samples.
  • Bags seal easily yet seal tightly
  • Approved by the FDA and USDA for food sampling
  • Composed of polyethylene blended with ethyl vinyl acetate for transparency and strength
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty