Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Economical Benchtop Refractometer, AR5, Reichert

Supplier: Reichert Technologies
14007000
The economical AR5 digital refractometer with Peltier temperature control included makes sophisticated measuring technology available to users on a tight budget

  • Reichert AR5 Advantages
  • Accurate, fast and reproducible measurements
  • Flexible, modular – easily connects to Reichert density meters, Density5 and Density4 and polarimeters, Polar2 and Polar3, for additional measurements
  • Robust design for rough environments
  • Sapphire prism
  • Built-in solid state Peltier thermostat for accurate temperature control
  • Easy to clean and maintain with flat sample chamber
  • Easy to calibrate
  • Energy saving LED light source
  • Remote diagnostics and maintenance of data available

Reichert AR Series Operating System Software—The operating system (OS) software has been designed with the user in mind. The intuitive touchscreen navigation saves time when defining settings or changing methods. The OS can easily connect the Reichert AR9 and AR5 to the Reichert density meters, Density5 and the Density4, and polarimeters, Polar2 and Polar3, making your lab ready for all applications. This powerful software makes the instrument part of your workflow communicating with LIMS systems and other laboratory instruments via USB or RS232. Programmed in the Universal Display, the software operates up to 5 measurement modules. Combine the OS with the optional, GxP software package to provide full compliance with 21 CFR Part 11 meeting all security electronic records requirements for the pharmaceutical industry. Reichert Technologies has created advanced optical and anlytical instruments for more that 150 years, and continues to pursue the highest level of accuracy and quality today. The AR5 is part of a modular system that delivers connectivity and flexibility to optimize your workflow.

High-end accuracy and a good price point give customers an exceptional advantage in the laboratory. The AR5 is precise, measuring refractive index and % Brix with results accurate to 0.0001 RI and 0.05% Brix. The AR5 is the ideal instrument to perform routine measurements with standard scales and has the ability to be configured for custom measurements. The robust design, sapphire prism and reliable operation make it a workhorse for the laboratory, resisting surface damage from demanding applications.

Certifications: RoHS, GMP/GLP. CE marked.

Ordering information: The Universal Display is an accessory for this product (sold separately).



 

ORDER

Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Economical Benchtop Refractomter, AR5 White/Stainless 14007000 10836-582 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Measurement Range (°Brix) 1.3200 - 1.5800RI/0 to 100% Brix
Scales Refractive index (RI), % Brix and other standard scales included.
Accuracy ± 0.0001 RI / ± 0.05% Brix
Reproducibility ± 0.0001 RI / ± 0.05% Brix
Temperature Compensation Range (°C) '+5° to +50°C (automatic)
Temperature Control Peltier control range 20°C or 25°C
Temperature Measurement Optimized accuracy – sensor integral to sample surface
Temperature Reproducibility ± 0.02°C
Temperature Accuracy ± 0.03°C
Interfaces RS232, USB optional
Light Source Light source - Lifetime LED, interference filter 589nm
Display Two line LCD
Measurement Mode Continuous
Ambient Operating Temperature +10 to +40°C
Measurement Method Single sample

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Color Touchscreen, Universal Display Operation system displayed on 7” color touchscreen. Drives up to 5 measurement modules. Program and save testing methods. White/stainless color. 14009500 10836-578 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty