Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultimate Benchtop Refractometer, AR9, Reichert

Supplier: Reichert Technologies
14009000
This top of the line AR9 refractometer is exceptionally precise with measurement accuracies of 0.00002 refractive index (RI) and 0.02% Brix

  • Reichert AR9 Advantages
  • Exceptional accuracy and reproducibility
  • Flexible, modular – easily connects to Reichert’s new Density5 and Density4 meters and Reichert’s new polarimeters, Polar2 and Polar3, for additional complimentary measurements
  • Continuous reading mode / real time measurement
  • Powerful software interface, compliant with critically regulated industries
  • YAG (Yttrium Aluminum Garnet) prism in stainless steel well, provides durable
  • maintenance-free optics
  • Built-in solid state Peltier thermostat for accurate temperature control
  • Modern, robust design is easy to clean and simplifies service
  • Sample temperatures up to 100°C
  • Easy to calibrate
  • Remote diagnostics and maintenance of data

Reichert AR Series Operating System Software—The operating system (OS) software has been designed with the user in mind. The intuitive touchscreen navigation saves time when defining settings or changing methods. The OS can easily connect the Reichert AR9 and AR5 to the Reichert density meters, Density5 and the Density4, and polarimeters, Polar2 and Polar3, making your lab ready for all applications. This powerful software makes the instrument part of your workflow communicating with LIMS systems and other laboratory instruments via USB or RS232. Programmed in the Universal Display, the software operates up to 5 measurement modules. Combine the OS with the optional, GxP software package to provide full compliance with 21 CFR Part 11 meeting all security electronic records requirements for the pharmaceutical industry. Reichert Technologies has created advanced optical and anlytical instruments for more that 150 years, and continues to pursue the highest level of accuracy and quality today. The AR9 is part of a modular system that delivers connectivity and flexibility to optimize your workflow.

With a built-in Peltier thermostat, the AR9 controls the temperature at the sample interface to an accuracy of 0.03°C. Backed by Reichert’s longstanding knowledge in the analytical instruments industry, the AR9 is the perfect match for research and development applications. This unit is resilient and durable, making it the right choice for all laboratory requirements. As a robust sealed system, the AR9 is able to perform in a variety of environments. This refractometer has the capability to provide measurements for the most critically regulated industries.  The AR9 has a YAG (Yttrium Aluminum Garnet) prism, the hardest prism available, guaranteeing accuracy and resilience for years. The AR9 includes our Universal Display which can be used to connect and control Density and Polarimeters as part of our modular system.

Certifications: RoHS, GMP/GLP. CE marked.

Ordering information: Universal Display included.



 

ORDER

Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Ultimate Benchtop Refractometer, AR9 White/Stainless 14009000 10836-580 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Measurement Range (°Brix) 1.32000 – 1.70000 RI / 0 to 100% Brix
Scales Refractive index (RI), %Brix and other standard scales included. User defined scales available.
Accuracy 1.32000 – 1.70000 RI / 0 to 100% Brix
Reproducibility ± 0.00001 RI / ± 0.01% Brix
Temperature Compensation Range (°C) '+5° to +100°C (automatic)
Temperature Control Peltier control range 5°C to +100°C
Temperature Measurement Optimized accuracy – sensor integral to sample surface
Temperature Reproducibility ± 0.02°C
Temperature Accuracy ± 0.03°C
Interfaces 4 USB, Mini USB, Ethernet, RS232
Light Source Light source - Lifetime LED, interference filter 589nm
Display Universal Display 7" color touchscreen
Measurement Mode Continuous
Ambient Operating Temperature +10 to +40°C
Measurement Method Single sample or flow through

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty