Would you like to log in again?
AntiBIND™ Microplates, Wheaton
Supplier: DWK Life Sciences
W266038 W266022 W266028 W266020 W266042 W266034 W266030 W266026 W266032 W266036 W266024 W266040
10852-156CS 478.21 USD InStock
10852-156 10852-152 75816-072 10852-150 10852-160 75816-078 75816-074 75816-070 75816-076 75816-080 10852-154 10852-158
AntiBIND™ Microplates, Wheaton
Microplates HTS Microplates
AntiBIND™ Microplates are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding
AntiBIND™ Microplates are manufactured in the United States with the highest purity polypropylene resin. They meet the SBS and ANSI dimensions for automation use. The conical shaped bottoms allow for maximum sample recovery and efficient mixing. All AntiBIND™ Microplates are tested and certified free of DNAase and RNase.
The treatment employs the use of Advanced Plasma Treatment (APT) technology to provide a comprehensive treatment without using any interfering leachables or additives that would migrate into solution during chromatography applications or long term storage. This results in increased protein recovery at low concentrations and more accurate and repeatable results. The microplates can be used in chromatography applications, long term sample storage, and bioanalytical assays.
Ordering information: AntiBIND™ Microplates are packaged 5 plates per pack.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty