Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Insterprep® Maximum Recovery Diluent, Seward

Supplier: Seward Laboratory System
INS-2101 INS-2301 INS-2201
Insterprep® is a new range of instantly soluble, sterile, granulated media specifically designed to speed up and simplify sample preparation for microbiological analysis

These instantly soluble, sterile Maximum Recovery Diluent media granules are supplied ready to use. Choose the Insterprep® sachet to fit specific protocol needs, or the Stomacher® bag. Simply add sterile distilled water and the sample as per protocol.

The Insterprep® pre-weighed, sterilized media bags are packaged with the correct amount of granular media, ready for instant use. The Insterprep® range of products is designed to help forward thinking laboratories cut waste and costs, manage laboratory resources more efficiently, and produce dependable and accurate results.




 

ORDER

Expand All / Collapse All
Description Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Maximum Recovery Diluent 225 mL Insterprep® Sachet INS-2201 10818-620 Pack of 100 Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Maximum Recovery Diluent 3375 mL Insterprep® Sachet INS-2301 10818-622 Pack of 100 Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Maximum Recovery Diluent 225 mL Insterprep® Stomacher® Bag INS-2101 10025-056 Pack of 100 (0.23.l) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty