Would you like to log in again?
DuPont™ Tyvek® IsoClean® Masks
Supplier: DuPont
Tyvek®
IC7310BU0003000B
10016-102CS 424.77 USD InStock
10016-102
DuPont™ Tyvek® IsoClean® Masks
Masks Cleanroom Masks
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound Tyvek® ties for cleanroom use
  • Pleated polyethylene outer
  • PBT filter
  • Metal nose piece
  • One size fits most
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty