Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)

Supplier: ACROS ORGANICS MS


AC61105-0040
200057-550
Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)


Formula: H₃CCH₂OH
MW: 46,07 g/mol
CAS Number: 64-17-5
UN: 1170
ADR: 3,III
Order Now
Specification Test Results
Molecular formula C2H6O
Linear formula C2H5OH
Flash point (°C) 12
CAS No 64-17-5
Version 0
Appearance (Form) Clear liquid
Identification passes test
Color scale =5 APHA
Assay 89 to 91 %
Refractive index 1.3580 to 1.3610 (25°C, 589 nm)
Water =0.10 %
Titrable acid =0.0003 meq/g
Titrable base =0.0002 meq/g
Residue after evaporation =5 ppm
KMnO4-reducing substance Passes test
UV at 205 nm A =1.00
UV at 210 nm A =0.65
UV at 220 nm A =0.35
UV at 254 nm A =0.04
Solubility (in water) Passes test
Methanol 4.0 to 5.0 %
Isopropyl alcohol 4.5 - 5.5%
Fluorescence background Passes test
Filtration test (truck) Passes test
LC use test Passes test


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty