Neivert Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1760
10194-268EA 236.66 USD
10194-268
Neivert Needle Holder, OR Grade, Sklar
Forceps
Neivert needle holder ensures a strong, anti-slip grip via its serrated jaws
Neivert needle holders are industry standard and constructed of stainless steel.
The angled ring handle eases comfort for the surgeon.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.