Reflex, Wound Clip Remover, 4", Roboz
Supplier: Roboz
RS-9263
100190-208EA 137.08 USD
100190-208
Reflex, Wound Clip Remover, 4", Roboz
Forceps
Reflex Wound Clip Remover; For Use With Reflex 9 mm And 7 mm Clips, And Other Brands; 4" Length