Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Babcock Forceps 6.25", 9 mm, Roboz Surgical

Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-8022
100189-912EA 168.46 USD InStock
100189-912
Babcock Forceps 6.25", 9 mm, Roboz Surgical
Forceps
Babcock Forceps; 9 mm Jaw Width; 6.25" Length

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty