Would you like to log in again?
Crile-Wood Needle Holder, Roboz Surgical
Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-7862
100189-052EA 134.31 USD InStock
100189-052
Crile-Wood Needle Holder, Roboz Surgical
Forceps
These holders are recommended for 4-0 to 6-0 suture.
Jaws are 2 to 4 mm wide
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty