Would you like to log in again?
Derf Needle Holder 4.75", Tungsten Carbide, Roboz Surgical
Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-7824
100190-066
Derf Needle Holder 4.75", Tungsten Carbide, Roboz Surgical
This Derf needle holder with carbide jaws is recommended for 1-0 to 4-0 suture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty