Would you like to log in again?
Schnidt Forceps,
7.5" Slight Curve, 1.7 mm tips, Roboz Surgical
Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-7341
100190-438
Schnidt Forceps,
7.5" Slight Curve, 1.7 mm tips, Roboz Surgical
Schnidt Forceps; Slight Curve; 7.5" Length
The tip width is 1.7 mm. The jaw width is 2.3 mm at 5 mm from the tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty