Jacobson Mosquito Forceps, 5", Curved, Extra Delicate, 0.9 mm tips, Roboz
Supplier: Roboz
RS-7117
100189-122EA 268.62 USD
100189-122
Jacobson Mosquito Forceps, 5", Curved, Extra Delicate, 0.9 mm tips, Roboz
Forceps
Jacobson Micro Mosquito Forceps; Extra Delicate; Light Weight; Curved; 5" Length
The tip width is 0.9 mm. The jaw width is 1.3 mm at 5 mm from the tip.