Halstead Mosquito Forceps, 5", Curved, 1.3 mm tips, Left Handed, Roboz
Supplier: Roboz
RS-7111L
100189-630EA 130.15 USD
100189-630
Halstead Mosquito Forceps, 5", Curved, 1.3 mm tips, Left Handed, Roboz
Forceps
Halstead Mosquito Forceps; Curved Left; 5" Length, 1.3 mm tip diameter, 2.1 mm jaw width
Left Handed