Vet Scissors 6.5" Hvy Straight Sharp/Blunt Serr, Roboz Surgical
Supplier: Roboz
RS-6940
100190-190EA 188.31 USD
100190-190
Vet Scissors 6.5" Hvy Straight Sharp/Blunt Serr, Roboz Surgical
Scissors
Veterinary Scissors; Heavy Pattern; Straight, Sharp-Blunt Serrated Blades; Blade Length 57 mm 6 1/2" Overall Length
Veterinary Scissors have one smooth blade and one serrated blade which allows for a sharp cut through softer bone and cartilage.