Mayo-Stille Scissors 5.5" Round Curved, Roboz Surgical
Supplier: Roboz
RS-6891
100189-220EA 114.92 USD
100189-220
Mayo-Stille Scissors 5.5" Round Curved, Roboz Surgical
Scissors
Mayo-Stille Scissors; Rounded, Curved Blades; 42 mm Blade Length ; 5 1/2" Overall Length