Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Micro Trephine 2.7 mm, Tungsten Carbide, Roboz Surgical

Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-6310
100188-828EA 83.08 USD InStock
100188-828
Micro Trephine 2.7 mm, Tungsten Carbide, Roboz Surgical
Dissection Sets
Micro Dissecting Trephine. Tungsten Carbide, 2.7 mm Diameter
 

The head diameter of these trephines is 2.7 mm. The shaft diameter is 2.33 mm. They work with our Ideal micro drill or other handpieces or drills accepting drill bits with 2.33 mm shaft diameter.

These items are designed for animal research only and are not intended for human use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty