Dissecting Micro Scissors, Straight, Roboz Surgical
Supplier: Roboz
RS-5680
100189-294EA 563.54 USD
100189-294
Dissecting Micro Scissors, Straight, Roboz Surgical
Scissors
Micro Dissecting Spring Scissors; Straight; 8 mm Cutting Edge; Flat Handle; 0.15 mm Tip Width; 6" Overall Length