Dissecting Spring Scissors, Castroviejo, Angled Blunt, Roboz Surgical
Supplier: Roboz
RS-5668
100189-454EA 456.92 USD
100189-454
Dissecting Spring Scissors, Castroviejo, Angled Blunt, Roboz Surgical
Scissors
Castroviejo Micro Dissecting Spring Scissors; Angular (Points To Left), Blunt Tips; 9 mm Cutting Edge; 0.15 mm Tip Width; 3 1/2" Overall Length