Dissecting Spring Scissors, Vannas, Angled On Edge, Roboz Surgical
Supplier: Roboz
RS-5618
100189-250EA 487.38 USD
100189-250
Dissecting Spring Scissors, Vannas, Angled On Edge, Roboz Surgical
Scissors
Vannas Micro Dissecting Spring Scissors; Angled On Edge (Points To Left When Laid Down); 3 mm Cutting Edge; 0.15 mm Tip Width; 3" Overall Length.