Would you like to log in again?
Dissecing Spring Scissors, Vannas, Curved, Roboz Surgical
Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-5611
100188-958
Dissecing Spring Scissors, Vannas, Curved, Roboz Surgical
Vannas Micro Dissecting Spring Scissors; Curved; 3 mm Cutting Edge; 0.15 mm Tip Width; 3" Overall Length  
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty