Dissecting Spring Scissors, McPherson-Vannas, Curved, Roboz Surgical
Supplier: Roboz
RS-5605
100190-414EA 517.85 USD
100190-414
Dissecting Spring Scissors, McPherson-Vannas, Curved, Roboz Surgical
Scissors
McPherson-Vannas Micro Dissecting Spring Scissors; Curved, Sharp Needle Points; 3 mm Cutting Edge; 0.1 mm Tip Width; 3" Overall Length.