Would you like to log in again?
Dissecing Spring Scissors, McPherson-Vannas, Curved, Roboz Surgical
Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-5601
100188-960EA 417.38 USD InStock
100188-960
Dissecing Spring Scissors, McPherson-Vannas, Curved, Roboz Surgical
Scissors
McPherson-Vannas Micro Dissecting Spring Scissors; Curved, Sharp Points; 5 mm Cutting Edge; 0.15 mm Tip Width; 3" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty