Micro Clip, Straight, 0.75×6 mm, Roboz
Supplier: Roboz
RS-5422
100188-878EA 113.54 USD
100188-878
Micro Clip, Straight, 0.75×6 mm, Roboz
Forceps
Micro Clip; Straight; 70 Grams Pressure; 0.75 mm Clip Width; 6 mm Jaw Length