Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Forceps, 6” (15 cm) Long Organ Holding Forceps, Roboz Surgical

Supplier: ROBOZ SURGICAL INSTR CO INC MS
RS-5255
100189-766EA 176.77 USD InStock
100189-766
Forceps, 6” (15 cm) Long Organ Holding Forceps, Roboz Surgical
Forceps
Singley Organ Holding Forceps; 6" Length.

The Singley heart holding forceps are designed to hold the heart of rats or other organs of similar size without being concerned about a sharp tip damaging the heart. A pin and screw allows you to adjust the opening of the tip to acco mmodate different size hearts. The pin and screw also holds the tip steady and in position to keep your hands free to conduct other steps of the procedure.

The Singley heart holding forceps are designed to hold the heart of rats or other organs of similar size without being concerned about a sharp tip damaging the heart

A pin and screw allows you to adjust the opening of the tip to acco mmodate different size hearts. The pin and screw also holds the tip steady and in position to keep your hands free to conduct other steps of the procedure.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty