Dumont #5 15 Deg Forceps, Inox, Tip Size 0.10×0.06 mm, Roboz
Supplier: Roboz
RS-5045A
100189-594EA 59.54 USD
100189-594
Dumont #5 15 Deg Forceps, Inox, Tip Size 0.10×0.06 mm, Roboz
Forceps
Dumont Tweezers; Pattern #5; 15 Degrees Up; .10 x .06 mm Tip Size; 105 mm Length; Material: Inox