Would you like to log in again?
m-EI Prepared Media Plates, Aquaplates
Supplier: AQUAPLATES, INC. MS
MEI12
102969-006PK 31.38 USD InStock
102969-006
m-EI Prepared Media Plates, Aquaplates
Media for Microbiology Prepared Media for Microbiology
For USEPA1600, m-EI prepared media plates for Enterococcus bacteria come in a convenient, resealable package
Designed for the enumeration of Enterococcus bacteria by Membrane Filtration.
m-EI plates are a single step verification for Enterococcus bacteria in surface, recreational, and waste waters. Each sleeve is date stamped with a 90 day expiration date. All orders come with a Quality Control Document referencing the ATCC (American Type Culture Colony) of each test organism as well as batch, sterility, and positive control information. Prepared media plates are designed for fast, consistent, and reliable results and are supplied ready to use. Each order comes with a full product guarantee as well as instructions for use. An NPDES Approved method.
Certifications: NPDES approved for reporting.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty