Would you like to log in again?
Modified m-TEC Prepared Media Plates, Aquaplates
Supplier: AQUAPLATES, INC. MS
MMTEC12
102969-002PK 22.15 USD InStock
102969-002
Modified m-TEC Prepared Media Plates, Aquaplates
Media for Microbiology Prepared Media for Microbiology
For USEPA 1603, Modified m-TEC prepared media plates for E
Designed for the enumeration of E. coli bacteria by Membrane Filtration.
coli come in a convenient, resealable package. Modified m-TEC plates are a single step verification for E. coli in surface, recreational, and waste waters. Each sleeve is date stamped with a 90 day expiration date. All orders come with a Quality Control Document referencing the ATCC (American Type Culture Colony) of each test organism, as well as sterility and positive control information. Prepared media plates are designed for fast, consistent, and reliable results and are supplied ready to use. Each order comes with a full product guarantee as well as instructions for use. An NPDES Approved method.
Certifications: NPDES approved for reporting.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty