Would you like to log in again?
VWR® Contour™ Deluxe FFAC Lab Chairs with Arms
Supplier: VWR International
VDAC-L-FFAC-R VDAC-H-FFAC-R VDAC-M-FFAC-R
10147-207EA 360.63 USD InStock
10147-207 10147-211 10147-209
VWR® Contour™ Deluxe FFAC Lab Chairs with Arms
Chairs
The VWR® Contour™ deluxe lab chairs with free-float articulating seat control and adjustable-height armrests with self-skinned urethane arms offer more ergonomic functions.
  • Pneumatic seat-height adjustment
  • Independent seat and backrest tilt
  • Black vinyl upholstery
The FFAC models feature pneumatic seat-height adjustment, backrest-height adjustment, and the seat and backrest tilt independently with separate levers and can free-float or lock into place. Look for standard black vinyl upholstery, a composite base, contoured-foam cushioned seats, and larger backrest with kidney support. Medium bench height and high bench height chairs also have an adjustable-height footring.
Certifications: Meet California Technical Bulletin 117 requirements.
Ordering information: VWR® Contour™ Lab Chairs ship within 72 hours of order; large quantity orders may require longer lead times.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty