Expression of C. elegans Kit
858
470136-492KT 275 USD
470136-492
Expression of C. elegans Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
Scientists can directly manipulate an organism's genome to produce a phenotype using engineered genes called transgenes. In this experiment, students will use fluorescent microscopy and PCR to analyze C. elegans (nematodes) that have been engineered to express the Green Fluorescent Protein (GFP).