Would you like to log in again?
PCR Quick Screening Kit Insertfinder, BioVision
Supplier: BioVision
K903-500
10013-826EA 211.15 USD InStock
10013-826
PCR Quick Screening Kit Insertfinder, BioVision
Nucleic Acid Purification Systems and Kits Plasmid Purification Systems and Kits
Directly screen colonies from plate
  • Designed to quickly screen large number of cloning inserts
Identification of clones that contain the desired DNA insert is an essential step in molecular cloning procedure. Traditional methods for identifying the correct DNA inserts require growing up individual colonies, isolating plasmid DNAs, and then digesting with restriction enzymes to analyze each individual clone. Such traditional procedures are very time consuming, laborious, and expensive. The new InsertFinderTM PCR Quick Screening Kit is designed to quickly screen clone candidates using PCR technology. The procedure allows screen of colonies directly from plate, without growing up the cultures. The proprietary Cell Lysis Buffer provided with the kit can efficiently lyse cells (bacteria or yeast) without interfering with PCR reactions. The PCR Enhancer included in the kit allows amplification of hairpin structure or GC-rich region efficiently. PCR products can directly be analyzed on an agarose gel without enzyme digestions. The kit provides a simple, convenient, and economical way to identify correct inserts. It is also suitable for screening large numbers of colonies.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty