Would you like to log in again?
Link-Fast 5 Minutes DNA Ligation Kit, BioVision
Supplier: BioVision
K902-50
10013-824EA 120.61 USD InStock
10013-824
Link-Fast 5 Minutes DNA Ligation Kit, BioVision
Ligation Kits
Ligates Sticky or Blunt ends within 5 min.
  • Provides unique formulations of reagents
  • For ligation of sticky-end or blunt-end DNA in just 5 minutes
The Link-FAST™ 5 minutes DNA Ligation Kit provides a fast and convenient means for ligation of sticky-end or blunt-end DNA fragments in just 5 minutes at room temperature. All necessary reagents required for ligation are provided. There is no need to prepare buffers or to add ATP or Mg²ᶧ. The kit can be used for cloning in plasmid vectors or phage vectors, linker ligation, recirculation of linear DNA, etc. Ligated DNA can be easily analyzed by agarose gel electrophoresis.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty