Would you like to log in again?
Gel-Fast Gel Staining/Destaining Kit, BioVision
Supplier: BioVision
K901-40
10013-822EA 114.73 USD InStock
10013-822
Gel-Fast Gel Staining/Destaining Kit, BioVision
Assays Enzyme Assays
Highly Sensitive, Stain/Destain within 20 min.
  • Gel Documentation machine, Naked eye
  • Provides a fast and sensitive method for SDS-PAGE gel staining and destaining
SDS-PAGE is one of the most powerful and commonly used techniques in molecular biology. However, traditional protein gel staining and destaining methods take several hours to finish and leaves a high blue background on the gel reducing the detection sensitivity. BioVision’s Gel-FASTTM 20 Minutes Gel Staining/Destaining Kit provides a fast and sensitive method for SDS-PAGE gel staining and destaining. The procedure requires only 5 minutes for gel staining and 15 minutes for destaining. A total of 20 minutes allow visualization of as little as 5 ng of protein on a water-clear background. The kit contains sufficient solutions to stain and destain up to 40 pieces of protein mini gels.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty