Would you like to log in again?
Dpp4 Inhibitor Screening Kit, BioVision
Supplier: BioVision
K780-100
10013-800
Dpp4 Inhibitor Screening Kit, BioVision
Simple Assay to Screen DPP4 Inhibitors, HTS
  • Fluorescence (Ex/Em 360/460 nm)
  • Can be used for screening of DPP4 inhibitors
Dipeptidyl peptidase-4 (DPP4) inhibitors have emerged as a new class of oral antidiabetic agents. These inhibitors promote glucose homeostasis by inhibiting degradation of glucose-dependent insulinotropic polypeptide and glucagon-like peptide-1 by DPP4. Glucagon-like peptide-1 extends the action of insulin while suppressing the release of glucagon. In BioVision’s DPP4 Inhibitor Screening Kit, DPP4 cleaves a substrate to release a quenched fluorescent group (Em/Ex = 360/460 nm). In presence of a DPP4 inhibitor, the cleavage will be inhibited. The kit provides a rapid, simple, sensitive, and reliable test, as well as, suitable for high throughput screening of DPP4 inhibitors. Sitagliptin is included as a control inhibitor to compare the efficacy of the test inhibitors.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty