Would you like to log in again?
Picoprobe G6PD Assay Kit Fluorometric, BioVision
Supplier: BioVision
K751-100
10013-300EA 515 USD InStock
10013-300
Picoprobe G6PD Assay Kit Fluorometric, BioVision
Assays Enzyme Assays
Glucose-6-Phosphate Dehydrogenase (G6PDH: EC 1.1.1.49) is a cytosolic enzyme in the pentose phosphate pathway.
  • Fluorescence (Ex/Em = 535/587 nm)
  • Measurement of G6PDH activity in various tissues and cells
  • Evaluation of pentose phosphate pathway
Glucose-6-Phosphate Dehydrogenase (G6PDH: EC 1.1.1.49) is a cytosolic enzyme in a metabolic pathway that supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). The NADPH in turn maintains the level of glutathione in these cells that helps protect the red blood cells against oxidative damage. Of greater quantitative importance is the production of NADPH for tissues actively engaged in biosynthesis of fatty acids and/or isoprenoids, such as liver, mammary gland, adipose tissue, and adrenal gland. BioVision’s PicoProbe™ Glucose-6-Phosphate Dehydrogenase Assay kit provides a quick and easy method for monitoring G6PDH activity in a wide variety of samples. In this assay, G6PDH converts G6P into pyruvate and NADPH, which further reduces PicoProbe™ to generate an intense fluorescence product (Ex/Em = 535/587 nm). This kit is simple, sensitive and high-throughput adaptable and can detect as low as 1 µU of G6PDH activity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty