Anti-CD185 Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L138D7]

Supplier: BioLegend
145521 145521 145522 145522
10756-484EA 386.31 USD
CA10756-484 CA10756-482 10756-484 10756-482
Anti-CD185 Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L138D7]
Antibodies
PE/Dazzle™ 594 anti-mouse CD185 (CXCR5) [L138D7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD185
Clonality: Monoclonal
Clone: L138D7
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR