Anti-ULK2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP3806A
89513-274EA 637.11 USD
89513-274
Anti-ULK2 Rabbit Polyclonal Antibody
Antibodies
Dot Blot: 1:500

Type: Primary
Antigen: ULK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR