Anti-CD185 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: L138D7]

Supplier: BioLegend
145513 145513
10759-190EA 280.11 USD
10759-190 CA10759-190
Anti-CD185 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: L138D7]
Antibodies
Brilliant Violet 605™ anti-mouse CD185 (CXCR5) [L138D7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD185
Clonality: Monoclonal
Clone: L138D7
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR