Anti-CD185 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: L138D7]

Supplier: BioLegend
145505 145505 145506 145506
10758-888EA 407.85 USD
CA10758-886 10758-888 10758-886 CA10758-888
Anti-CD185 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: L138D7]
Antibodies
APC anti-mouse CD185 (CXCR5) [L138D7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD185
Clonality: Monoclonal
Clone: L138D7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR