Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD185 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: J252D4]

Supplier: Biolegend

356912 356912 356911 356911
CA10760-210 10760-210 CA10760-208 10760-208
Anti-CD185 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: J252D4]
Antibodies
Alexa Fluor® 488 anti-human CD185 (CXCR5) [J252D4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD185
Clonality: Monoclonal
Clone: J252D4
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty