Anti-IL6RA Mouse Monoclonal Antibody [clone: B-N12]

Supplier: Cell Sciences
CDM010
102995-332EA 540 USD
102995-332
Anti-IL6RA Mouse Monoclonal Antibody [clone: B-N12]
Antibodies
No effect on IL-6 induced proliferation of XG-1 cell line

Type: Primary
Antigen: IL6RA
Clonality: Monoclonal
Clone: B-N12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR