Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD184 Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L276F12]

Supplier: Biolegend

146513 146514 146513 146514
CA10759-200 CA10759-202 10759-200 10759-202
Anti-CD184 Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L276F12]
Antibodies
PE/Dazzle™ 594 anti-mouse CD184 (CXCR4) [L276F12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD184
Clonality: Monoclonal
Clone: L276F12
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty