Anti-CD184 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: L276F12]

Supplier: BioLegend
146508 146508 146507 146507
10759-192EA 392.16 USD
10759-192 CA10759-192 CA10756-550 10756-550
Anti-CD184 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: L276F12]
Antibodies
APC anti-mouse CD184 (CXCR4) [L276F12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD184
Clonality: Monoclonal
Clone: L276F12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR