Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD184 Rat Monoclonal Antibody [clone: L276F12]

Supplier: Biolegend

146502 146502
10756-540EA 200.92 USD InStock
10756-540 CA10756-540
Anti-CD184 Rat Monoclonal Antibody [clone: L276F12]
Antibodies
Purified anti-mouse CD184 (CXCR4) [L276F12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD184
Clonality: Monoclonal
Clone: L276F12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty