Anti-CD183 (CXCR3) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 650) [clone: CXCR3-173]

Supplier: BioLegend
126531 126531
10757-056EA 327.32 USD
CA10757-056 10757-056
Anti-CD183 (CXCR3) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 650) [clone: CXCR3-173]
Antibodies
Brilliant Violet 650™ anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD183 (CXCR3)
Clonality: Monoclonal
Clone: CXCR3-173
Conjugation: Brilliant Violet® 650
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR